Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342626.1 | NP_001331798.1 | 3 - 4 | 1191 | 77 | |
NM_120270.4 | NP_195812.1 | 3 - 4 | 1191 | 77 | ||
GenBank mRNA | AK117628.1 | - | 3 - 4 | 1166 | 77 | |
BT006132.1 | - | 3 - 4 | 1144 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |