Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120325.4 | NP_195867.2 | 8 - 9 | 966 | 120 | |
NM_180696.2 | NP_851027.1 | 9 - 10 | 775 | 120 | ||
NM_180697.3 | NP_851028.1 | 9 - 10 | 921 | 120 | ||
GenBank mRNA | AJ294531.1 | - | 9 - 10 | 847 | 120 | |
AJ319027.1 | - | 8 - 9 | 752 | 120 | ||
AK117135.1 | - | 9 - 10 | 737 | 120 | ||
BT005286.1 | - | 8 - 9 | 674 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |