Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001342672.1 | NP_001332134.1 | 7 - 8 | 1160 | 69 | |
NM_120362.4 | NP_568108.1 | 7 - 8 | 972 | 69 | ||
NM_180425.2 | NP_850756.1 | 7 - 8 | 914 | 69 | ||
GenBank mRNA | AJ937209.1 | - | 7 - 8 | 826 | 69 | |
AY519514.1 | - | 7 - 8 | 826 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |