Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO70A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125690.1 | NP_001119162.1 | 9 - 10 | 1816 | 86 | |
NM_001203287.1 | NP_001190216.1 | 11 - 12 | 1774 | 86 | ||
NM_120434.5 | NP_195974.2 | 10 - 11 | 1877 | 86 | ||
GenBank mRNA | AY072155.1 | - | 10 - 11 | 1697 | 86 | |
AY133751.1 | - | 10 - 11 | 1656 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |