Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EXO70A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125690.1 | NP_001119162.1 | 6 - 7 | 1414 | 112 | |
NM_001203287.1 | NP_001190216.1 | 8 - 9 | 1372 | 112 | ||
NM_120434.5 | NP_195974.2 | 7 - 8 | 1475 | 112 | ||
GenBank mRNA | AY072155.1 | - | 7 - 8 | 1295 | 112 | |
AY133751.1 | - | 7 - 8 | 1254 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |