Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IREG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203288.1 | NP_001190217.1 | 3 - 4 | 323 | 101 | |
NM_001342716.1 | NP_001332410.1 | 2 - 3 | 491 | 101 | ||
NM_120438.4 | NP_195977.2 | 2 - 3 | 493 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |