Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120465.3 | NP_196004.1 | 3 - 4 | 388 | 60 | |
GenBank mRNA | BT024828.1 | - | 3 - 4 | 312 | 60 | |
U77674.1 | - | 3 - 4 | 327 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |