Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDH1-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127401.3 | NP_179435.1 | 5 - 6 | 671 | 108 | |
GenBank mRNA | AK119142.1 | - | 5 - 6 | 650 | 108 | |
BT005938.1 | - | 5 - 6 | 647 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |