Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127427.3 | NP_179461.1 | 1 - 2 | 234 | 122 | |
GenBank mRNA | AF424550.1 | - | 1 - 2 | 219 | 122 | |
AY058103.1 | - | 1 - 2 | 219 | 122 | ||
AY065093.1 | - | 1 - 2 | 226 | 122 | ||
BT002632.1 | - | 1 - 2 | 201 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |