Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127427.3 | NP_179461.1 | 5 - 6 | 1458 | 66 | |
GenBank mRNA | AF424550.1 | - | 5 - 6 | 1443 | 66 | |
AY058103.1 | - | 5 - 6 | 1443 | 66 | ||
AY065093.1 | - | 5 - 6 | 1450 | 66 | ||
BT002632.1 | - | 5 - 6 | 1425 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |