Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127464.2 | NP_179497.1 | 1 - 2 | 994 | 60 | |
GenBank mRNA | BT002975.1 | - | 1 - 2 | 994 | 60 | |
BT004456.1 | - | 1 - 2 | 967 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |