Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335621.1 | NP_001318247.1 | 7 - 8 | 1542 | 108 | |
NM_001335622.1 | NP_001324425.1 | 8 - 9 | 1894 | 108 | ||
NM_127468.5 | NP_179501.1 | 8 - 9 | 1696 | 108 | ||
GenBank mRNA | AJ297264.1 | - | 7 - 8 | 1501 | 108 | |
AK227102.1 | - | 8 - 9 | 1605 | 108 | ||
AY096796.1 | - | 8 - 9 | 1602 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |