Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ckl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127505.5 | NP_179537.1 | 13 - 14 | 1385 | 85 | |
GenBank mRNA | AY086262.1 | - | 13 - 14 | 1317 | 85 | |
AY099837.1 | - | 13 - 14 | 1276 | 85 | ||
AY943844.1 | - | 13 - 14 | 1078 | 85 | ||
BT006580.1 | - | 13 - 14 | 1078 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |