Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CKX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127508.2 | NP_565455.1 | 2 - 3 | 746 | 104 | |
GenBank mRNA | BT004107.1 | - | 2 - 3 | 746 | 104 | |
BT005653.1 | - | 2 - 3 | 692 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Arabidopsis thaliana Probe spans exons |