Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EER5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335638.1 | NP_001324728.1 | 8 - 9 | 654 | 77 | |
NM_127514.4 | NP_179546.2 | 10 - 11 | 1006 | 77 | ||
GenBank mRNA | AK118897.1 | - | 10 - 11 | 977 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |