Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EER5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335638.1 | NP_001324728.1 | 3 - 4 | 284 | 124 | |
NM_127514.4 | NP_179546.2 | 5 - 6 | 636 | 124 | ||
GenBank mRNA | AK118897.1 | - | 5 - 6 | 607 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |