Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127525.5 | NP_179557.1 | 6 - 7 | 808 | 127 | |
GenBank mRNA | AK227449.1 | - | 6 - 7 | 664 | 127 | |
BT006491.1 | - | 6 - 7 | 628 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |