Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: rps15ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127530.5 | NP_179562.1 | 1 - 2 | 266 | 83 | |
GenBank mRNA | BT014766.1 | - | 1 - 2 | 188 | 83 | |
BT014999.1 | - | 1 - 2 | 129 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |