Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127538.4 | NP_565459.1 | 4 - 5 | 889 | 102 | |
GenBank mRNA | BT033135.1 | - | 4 - 5 | 464 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |