Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001335672.1 | NP_001325368.1 | 17 - 18 | 2176 | 66 | |
NM_001335673.1 | NP_001325369.1 | 17 - 18 | 2176 | 66 | ||
NM_001335674.1 | NP_001325367.1 | 17 - 18 | 2183 | 66 | ||
NM_127588.3 | NP_179619.2 | 17 - 18 | 1985 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |