Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHB6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084455.1 | NP_001077924.1 | 5 - 6 | 595 | 76 | |
NM_127613.3 | NP_179643.1 | 4 - 5 | 546 | 76 | ||
GenBank mRNA | AY072378.1 | - | 4 - 5 | 527 | 76 | |
AY088363.1 | - | 4 - 5 | 544 | 76 | ||
AY114620.1 | - | 3 - 4 | 455 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |