Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202633.1 | NP_001189562.1 | 3 - 4 | 775 | 75 | |
NM_127617.3 | NP_565476.1 | 2 - 3 | 783 | 75 | ||
GenBank mRNA | AB062489.1 | - | 2 - 3 | 727 | 75 | |
AY026772.1 | - | 2 - 3 | 599 | 75 | ||
AY054600.1 | - | 2 - 3 | 673 | 75 | ||
AY081485.1 | - | 2 - 3 | 599 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |