Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FIO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036305.1 | NP_001031382.1 | 1 - 2 | 308 | 68 | |
NM_001202638.2 | NP_001189567.1 | 1 - 2 | 319 | 68 | ||
NM_127677.3 | NP_179703.2 | 1 - 2 | 308 | 68 | ||
GenBank mRNA | BT015875.1 | - | 1 - 2 | 288 | 68 | |
BT020210.1 | - | 1 - 2 | 231 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |