Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127685.4 | NP_179711.2 | 10 - 11 | 1128 | 70 | |
GenBank mRNA | BT000912.1 | - | 10 - 11 | 885 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |