Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BPC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_127696.4 | NP_565503.1 | 2 - 3 | 347 | 118 | |
NM_179681.2 | NP_850012.1 | 2 - 3 | 162 | 118 | ||
GenBank mRNA | AK229090.1 | - | 2 - 3 | 128 | 118 | |
AK317165.1 | - | 2 - 3 | 198 | 118 | ||
AY084554.1 | - | 2 - 3 | 252 | 118 | ||
BT026109.1 | - | 1 - 2 | 64 | 118 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |