Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UFD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001036307.2 | NP_001031384.1 | 8 - 9 | 1408 | 81 | |
NM_001084464.1 | NP_001077933.1 | 9 - 10 | 1200 | 81 | ||
NM_001335738.1 | NP_001323742.1 | 9 - 10 | 1264 | 81 | ||
NM_001335739.1 | NP_001323741.1 | 9 - 10 | 1183 | 81 | ||
NM_127699.5 | NP_565504.1 | 9 - 10 | 964 | 81 | ||
GenBank mRNA | AK226526.1 | - | 9 - 10 | 965 | 81 | |
AK317375.1 | - | 9 - 10 | 1201 | 81 | ||
BT033134.1 | - | 7 - 8 | 862 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |