Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338816.1 | NP_001319649.1 | 5 - 6 | 1279 | 85 | |
NM_001338817.1 | NP_001319650.1 | 5 - 6 | 1279 | 85 | ||
NM_001338818.1 | NP_001327585.1 | 5 - 6 | 1256 | 85 | ||
NM_180304.2 | NP_850635.1 | 5 - 6 | 1297 | 85 | ||
GenBank mRNA | AK117422.1 | - | 5 - 6 | 1157 | 85 | |
BT005000.1 | - | 5 - 6 | 1157 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |