Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDX13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113580.2 | NP_189303.1 | 3 - 4 | 514 | 141 | |
NM_180305.2 | NP_850636.1 | 2 - 3 | 578 | 141 | ||
GenBank mRNA | BT021980.1 | - | 3 - 4 | 344 | 141 | |
BT024767.1 | - | 2 - 3 | 195 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |