Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDU6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_180319.2 | NP_850650.1 | 1 | 310 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |