Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SETH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001339193.1 | NP_001326667.1 | 3 - 4 | 937 | 77 | |
NM_114379.3 | NP_566874.1 | 3 - 4 | 791 | 77 | ||
NM_180327.2 | NP_850658.1 | 3 - 4 | 791 | 77 | ||
GenBank mRNA | AY039602.1 | - | 3 - 4 | 788 | 77 | |
AY081726.1 | - | 2 - 3 | 691 | 77 | ||
KJ138805.1 | - | 2 - 3 | 691 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |