Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114613.3 | NP_190329.2 | 12 - 13 | 1538 | 109 | |
NM_180335.1 | NP_850666.1 | 12 - 13 | 1538 | 109 | ||
GenBank mRNA | AK226195.1 | - | 12 - 13 | 1543 | 109 | |
BT015353.1 | - | 12 - 13 | 1443 | 109 | ||
BT015887.1 | - | 12 - 13 | 1440 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |