Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114613.3 | NP_190329.2 | 6 - 7 | 954 | 80 | |
NM_180335.1 | NP_850666.1 | 6 - 7 | 954 | 80 | ||
GenBank mRNA | AK226195.1 | - | 6 - 7 | 959 | 80 | |
BT015353.1 | - | 6 - 7 | 859 | 80 | ||
BT015887.1 | - | 6 - 7 | 856 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |