Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114680.4 | NP_190394.3 | 3 - 4 | 704 | 101 | |
GenBank mRNA | AY136413.1 | - | 3 - 4 | 620 | 101 | |
BT012191.1 | - | 3 - 4 | 563 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |