Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114680.4 | NP_190394.3 | 6 - 7 | 931 | 140 | |
GenBank mRNA | AY136413.1 | - | 6 - 7 | 847 | 140 | |
BT012191.1 | - | 6 - 7 | 790 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |