Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U1-70K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_114927.4 | NP_190636.1 | 3 - 4 | 484 | 75 | |
NM_180345.1 | NP_850676.1 | 3 - 4 | 376 | 75 | ||
GenBank mRNA | AY039874.1 | - | 3 - 4 | 349 | 75 | |
AY094003.1 | - | 3 - 4 | 292 | 75 | ||
M93439.1 | - | 3 - 4 | 315 | 75 | ||
U52910.1 | - | 3 - 4 | 376 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |