Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UEV1D-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084803.1 | NP_001078272.1 | 3 - 4 | 452 | 65 | |
NM_001203144.1 | NP_001190073.1 | 3 - 4 | 491 | 65 | ||
NM_115116.4 | NP_566968.1 | 3 - 4 | 489 | 65 | ||
NM_180353.2 | NP_850684.1 | 3 - 4 | 429 | 65 | ||
GenBank mRNA | AY042846.1 | - | 3 - 4 | 371 | 65 | |
AY072492.1 | - | 3 - 4 | 296 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |