Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001084809.2 | NP_001078278.1 | 1 - 2 | 598 | 93 | |
NM_115221.4 | NP_190929.1 | 2 - 3 | 475 | 93 | ||
NM_180365.3 | NP_850696.1 | 2 - 3 | 148 | 93 | ||
GenBank mRNA | AK228083.1 | - | 2 - 3 | 175 | 93 | |
AK230120.1 | - | 586 | 93 | |||
AY086756.1 | - | 2 - 3 | 185 | 93 | ||
BT004779.1 | - | 1 - 2 | 70 | 93 | ||
U82434.1 | - | 2 - 3 | 90 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |