Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115261.4 | NP_566993.1 | 7 - 8 | 758 | 75 | |
NM_180370.3 | NP_850701.1 | 6 - 7 | 788 | 75 | ||
GenBank mRNA | AY056265.1 | - | 7 - 8 | 690 | 75 | |
AY096635.1 | - | 7 - 8 | 537 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |