Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115798.3 | NP_567083.1 | 12 - 13 | 1444 | 80 | |
NM_180390.3 | NP_850721.1 | 12 - 13 | 1565 | 80 | ||
GenBank mRNA | AF360241.1 | - | 12 - 13 | 1430 | 80 | |
AY040039.1 | - | 11 - 12 | 1116 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |