Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115798.3 | NP_567083.1 | 3 - 4 | 612 | 150 | |
NM_180390.3 | NP_850721.1 | 3 - 4 | 733 | 150 | ||
GenBank mRNA | AF360241.1 | - | 3 - 4 | 598 | 150 | |
AY040039.1 | - | 2 - 3 | 284 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |