Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_115798.3 | NP_567083.1 | 8 - 9 | 1045 | 148 | |
NM_180390.3 | NP_850721.1 | 8 - 9 | 1166 | 148 | ||
GenBank mRNA | AF360241.1 | - | 8 - 9 | 1031 | 148 | |
AY040039.1 | - | 7 - 8 | 717 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |