Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_116025.4 | NP_567115.1 | 3 - 4 | 803 | 64 | |
NM_180402.2 | NP_850733.1 | 3 - 4 | 803 | 64 | ||
GenBank mRNA | AF292397.1 | - | 3 - 4 | 597 | 64 | |
BT030347.1 | - | 3 - 4 | 521 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |