Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203285.1 | NP_001190214.1 | 2 - 3 | 707 | 61 | |
NM_180426.3 | NP_850757.1 | 2 - 3 | 508 | 61 | ||
GenBank mRNA | AB493733.1 | - | 1 - 2 | 248 | 61 | |
AJ294532.1 | - | 1 - 2 | 248 | 61 | ||
AK228148.1 | - | 2 - 3 | 489 | 61 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |