Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_120525.4 | NP_196063.1 | 3 - 4 | 613 | 77 | |
NM_180433.3 | NP_850764.1 | 3 - 4 | 613 | 77 | ||
GenBank mRNA | AF375411.1 | - | 3 - 4 | 475 | 77 | |
AY086553.1 | - | 3 - 4 | 493 | 77 | ||
BT000854.1 | - | 2 - 3 | 324 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |