Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDF1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343352.1 | NP_001332718.1 | 5 - 6 | 794 | 112 | |
NM_121470.3 | NP_196970.1 | 5 - 6 | 794 | 112 | ||
NM_180490.1 | NP_850821.1 | 5 - 6 | 794 | 112 | ||
GenBank mRNA | AY050879.1 | - | 5 - 6 | 794 | 112 | |
AY096673.1 | - | 5 - 6 | 719 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |