Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001343401.1 | NP_001318568.1 | 1 - 2 | 947 | 79 | |
NM_121550.3 | NP_568315.1 | 2 - 3 | 281 | 79 | ||
NM_180493.4 | NP_850824.1 | 2 - 3 | 611 | 79 | ||
GenBank mRNA | AK228599.1 | - | 2 - 3 | 281 | 79 | |
AY086424.1 | - | 2 - 3 | 275 | 79 | ||
BT005773.1 | - | 2 - 3 | 281 | 79 | ||
BT006108.1 | - | 1 - 2 | 126 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Arabidopsis thaliana Probe spans exons |