Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121620.2 | NP_568328.1 | 4 - 5 | 536 | 98 | |
NM_180497.2 | NP_850828.1 | 4 - 5 | 621 | 98 | ||
NM_203058.3 | NP_974787.1 | 3 - 4 | 708 | 98 | ||
GenBank mRNA | AY058152.1 | - | 4 - 5 | 536 | 98 | |
AY091052.1 | - | 4 - 5 | 520 | 98 | ||
AY117359.1 | - | 3 - 4 | 434 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |