Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGLCT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121620.2 | NP_568328.1 | 6 - 7 | 778 | 137 | |
NM_180497.2 | NP_850828.1 | 6 - 7 | 863 | 137 | ||
NM_203058.3 | NP_974787.1 | 5 - 6 | 950 | 137 | ||
GenBank mRNA | AY058152.1 | - | 6 - 7 | 778 | 137 | |
AY091052.1 | - | 6 - 7 | 762 | 137 | ||
AY117359.1 | - | 5 - 6 | 676 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |