Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_121634.4 | NP_197133.1 | 10 - 11 | 1405 | 93 | |
NM_180498.3 | NP_850829.1 | 10 - 11 | 1392 | 93 | ||
GenBank mRNA | AK226906.1 | - | 10 - 11 | 1331 | 93 | |
BT021989.1 | - | 10 - 11 | 1294 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |