Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203404.1 | NP_001190333.1 | 7 - 8 | 1514 | 91 | |
NM_121888.3 | NP_197384.1 | 7 - 8 | 1609 | 91 | ||
NM_180519.1 | NP_850850.1 | 7 - 8 | 1482 | 91 | ||
GenBank mRNA | AF367355.1 | - | 7 - 8 | 1450 | 91 | |
AJ011612.1 | - | 7 - 8 | 1408 | 91 | ||
AY063815.1 | - | 7 - 8 | 1482 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |